Learn More
Description
Specifications
Specifications
| Antigen | TREM1 |
| Applications | Western Blot, KnockDown |
| Classification | Monoclonal |
| Clone | CL7111 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 μg/mL, Knockdown Validated |
| Formulation | PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide |
| Gene Alias | CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 |
| Gene Symbols | TREM1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
