Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREM1 Antibody (CL7111), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | TREM1 |
---|---|
Clone | CL7111 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Host Species | Mouse |
Description
TREM1 Monoclonal specifically detects TREM1 in Human samples. It is validated for Western Blot, Knockdown Validated.Specifications
TREM1 | |
Monoclonal | |
Mouse | |
PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
54210 | |
This antibody was developed against a synthetic peptide corresponding to amino acids: KLTEEKYELKEGQTLDVK | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
CL7111 | |
Unconjugated | |
Human | |
CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 | |
TREM1 | |
IgG1 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title