Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREM1 Antibody (L5-B8.2A12.3A12) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018512 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018512 0.05 mg
NBP140907 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018512 Supplier Novus Biologicals Supplier No. NBP1409070.05MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TREM1 Monoclonal specifically detects TREM1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, CyTOF-ready.

Specifications

Antigen TREM1
Applications CyTOF, Immunofluorescence, Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone L5-B8.2A12.3A12
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:25-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready
Formulation Phosphate buffered saline with No Preservative
Gene Accession No. Q9JKE2
Gene Alias CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1
Gene Symbols TREM1
Host Species Rat
Immunogen Mouse TREM-1 Fc fusion protein.
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54210
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.