Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREM1 Antibody (L5-B8.2A12.3A12) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $560.50
Specifications
| Antigen | TREM1 |
|---|---|
| Clone | L5-B8.2A12.3A12 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:25-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready |
| Applications | CyTOF, Immunofluorescence, Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen), Immunocytochemistry |
Description
TREM1 Monoclonal specifically detects TREM1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, CyTOF-ready.Specifications
| TREM1 | |
| 1.0 mg/mL | |
| CyTOF, Immunofluorescence, Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen), Immunocytochemistry | |
| Unconjugated | |
| Rat | |
| Signal Transduction | |
| Q9JKE2 | |
| 54210 | |
| Mouse TREM-1 Fc fusion protein. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| L5-B8.2A12.3A12 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:25-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1 | |
| TREM1 | |
| IgG2a | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title