Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREM1 Antibody (MM0583-9T3), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP211977IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211977IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211977IR Supplier Novus Biologicals Supplier No. NBP211977IR

Mouse Monoclonal Antibody

TREM1 Monoclonal specifically detects TREM1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TREM1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0583-9T3
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Gene Accession No. Q9NP99
Gene Alias CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1
Gene Symbols TREM1
Host Species Mouse
Immunogen Human recombinant TREM1 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54210
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.