Learn More
Description
Specifications
Specifications
| Antigen | TREM2 |
| Applications | ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 14 |
| Conjugate | Alexa Fluor 700 |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TREM2 (NP_112544.1; Met1-Pro168). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
