Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREM2 Antibody (08), FITC, Novus Biologicals™
SDP

Catalog No. NB277899 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB277899 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB277899 Supplier Novus Biologicals Supplier No. NBP321453F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TREM2 Monoclonal antibody specifically detects TREM2 in Human samples. It is validated for ELISA, ELISA (Matched Antibody Pair)

Specifications

Antigen TREM2
Applications ELISA
Classification Monoclonal
Clone 08
Conjugate FITC
Dilution ELISA, ELISA Detection (Matched Antibody Pair)
Formulation PBS
Gene Alias Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2
Host Species Mouse
Immunogen Purified, recombinant Human TREM2 (NBP3-21453PEP; NP_061838.1; Met1-Ser174)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Immunology, Inflammation, Innate Immunity, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 54209
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.