Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREM2 Antibody (237920R), FITC, Novus Biologicals™
SDP

Catalog No. NB297039 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297039 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297039 Supplier Novus Biologicals Supplier No. FAB17291RF
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TREM2 Monoclonal specifically detects TREM2 in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen TREM2
Applications Flow Cytometry
Classification Monoclonal
Clone 237920R
Conjugate FITC
Dilution Flow Cytometry
Formulation PBS
Gene Alias Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TREM2, Met1-Pro168, Accession # Q99NH8
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Immunology, Inflammation, Innate Immunity, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 54209
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.