Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREM2 Antibody (2B5), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N107101PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N107101PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N107101PEC7 Supplier Novus Biologicals Supplier No. NBP107101PECY7

Mouse Monoclonal Antibody

TREM2 Monoclonal specifically detects TREM2 in Human, Rat samples. It is validated for Flow Cytometry, Product Image.

Specifications

Antigen TREM2
Applications Flow Cytometry
Classification Monoclonal
Clone 2B5
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2
Gene Symbols TREM2
Host Species Mouse
Immunogen Recombinant human TREM2 (19-161aa) purified from E. coli (NP_061838).
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Alzheimers Research, Immunology, Inflammation, Innate Immunity, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 54209.0
Target Species Human, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.