Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TREM2 Monoclonal Antibody (08), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA543382
This item is not returnable.
View return policy
Description
Specificity: Human TREM-2/TREM2
TREM2 is a membrane protein that forms a receptor signaling complex with TYROBP. The encoded protein may be involved in chronic inflammation by triggering the production of constitutive inflammatory cytokines. Defects in this gene are a cause of polycystic lipomembranous osteodysplasia with sclerosing leukoencephalopathy (PLOSL).
Specifications
TREM2 | |
Monoclonal | |
1 mg/mL | |
PBS with 10% trehalose and no preservative; pH 7.0 | |
Q9NZC2 | |
Trem2 | |
Recombinant Human TREM-2/TREM2 Protein. | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
08 | |
Unconjugated | |
Trem2 | |
OTTHUMP00000039771; soluble TREM 2; soluble TREM2; sTREM 2; sTREM2; TREM; Trem2; TREM-2; Trem2a; Trem2b; Trem2c; Triggering receptor expressed on monocytes 2; Triggering receptor expressed on myeloid cells; triggering receptor expressed on myeloid cells 2; triggering receptor expressed on myeloid cells 2a; triggering receptor expressed on myeloid cells 2b; triggering receptor expressed on myeloid cells 2c | |
Mouse | |
Protein A | |
RUO | |
54209 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction