Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREM2 Monoclonal Antibody (08), Invitrogen™
Mouse Monoclonal Antibody
$285.50 - $493.00
Specifications
Antigen | TREM2 |
---|---|
Clone | 08 |
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543382
|
Invitrogen™
MA543382 |
50 μL |
Each for $285.50
|
|
|||||
PIMA543015
|
Invitrogen™
MA543015 |
100 μL |
Each for $493.00
|
|
|||||
Description
Specificity: Human TREM-2/TREM2
TREM2 is a membrane protein that forms a receptor signaling complex with TYROBP. The encoded protein may be involved in chronic inflammation by triggering the production of constitutive inflammatory cytokines. Defects in this gene are a cause of polycystic lipomembranous osteodysplasia with sclerosing leukoencephalopathy (PLOSL).Specifications
TREM2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 10% trehalose and no preservative; pH 7.0 | |
OTTHUMP00000039771; soluble TREM 2; soluble TREM2; sTREM 2; sTREM2; TREM; Trem2; TREM-2; Trem2a; Trem2b; Trem2c; Triggering receptor expressed on monocytes 2; Triggering receptor expressed on myeloid cells; triggering receptor expressed on myeloid cells 2; triggering receptor expressed on myeloid cells 2a; triggering receptor expressed on myeloid cells 2b; triggering receptor expressed on myeloid cells 2c | |
Trem2 | |
IgG1 | |
Protein A | |
Antibody |
08 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
Q9NZC2 | |
54209 | |
Recombinant Human TREM-2/TREM2 Protein. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Trem2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title