Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREM2b Antibody (RM0139-5J46) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP122515
Description
TREM2b Monoclonal specifically detects TREM2b in Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| TREM2b | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1:1000, Flow Cytometry, Immunohistochemistry, Immunocytochemistry 1:50-1:100 | |
| Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 54209 | |
| Reconstitute with sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
| RM0139-5J46 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
| TREM2 | |
| Mouse recombinant TREM2b (extracellular domain) | |
| 0.1 mg | |
| Primary | |
| This antibody detects mouse of TREM-2b, but not TREM-1 in Western blotting. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction