Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TREM2b Antibody (RM0139-5J46) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50
Specifications
Antigen | TREM2b |
---|---|
Clone | RM0139-5J46 |
Concentration | LYOPH |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
TREM2b Monoclonal specifically detects TREM2b in Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
TREM2b | |
LYOPH | |
Unconjugated | |
Rat | |
Mouse | |
54209 | |
IgG2 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
RM0139-5J46 | |
Monoclonal | |
Purified | |
RUO | |
Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
TREM2 | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title