Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREML1/TLT-1 Antibody (268420), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB365367 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB365367 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB365367 Supplier Novus Biologicals Supplier No. FAB2394A
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TREML1/TLT-1 Monoclonal specifically detects TREML1/TLT-1 in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen TREML1/TLT-1
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 268420
Conjugate APC
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias dJ238O23.3, GLTL1825, PRO3438, TLT-1, TLT1MGC119173, trem-like transcript 1 protein, triggering receptor expressed on myeloid cells-like 1, Triggering receptor expressed on myeloid cells-like protein 1
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human TREML1/TLT-1, Gln16-Ser160, Accession # Q86YW5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340205
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.