Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREML2/TLT-2 Antibody (656906), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB311544 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311544 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311544 Supplier Novus Biologicals Supplier No. FAB6707APCCY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TREML2/TLT-2 Monoclonal antibody specifically detects TREML2/TLT-2 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen TREML2/TLT-2
Applications Flow Cytometry
Classification Monoclonal
Clone 656906
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias C6orf76, dJ238O23.1, FLJ13693, MGC149715, MGC149716, TLT2, TLT-2, trem-like transcript 2 protein, triggering receptor expressed on myeloid cells-like 2, triggering receptor expressed on myeloid cells-like protein 2, UNQ6268/PRO20473
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse TREML2/TLT-2, His25-Ala270, Accession # Q2LA85
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79865
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.