Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREML4/TLT-4 Antibody (817914), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB377895 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB377895 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB377895 Supplier Novus Biologicals Supplier No. FAB5500J
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TREML4/TLT-4 Monoclonal antibody specifically detects TREML4/TLT-4 in Mouse samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen TREML4/TLT-4
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 817914
Conjugate Janelia Fluor 646
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias TLT4
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TREML4/TLT-4, Gly25-Pro195, Accession # Q3LRV9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285852
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.