Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRERF1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10081655 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10081655 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10081655 Supplier Novus Biologicals Supplier No. NB10081655
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRERF1 Polyclonal specifically detects TRERF1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen TRERF1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:2000 to 1:10000
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. Q96PN7
Gene Alias BCAR2, Breast cancer estrogen resistance 2RAPAHSA277276, dJ139D8.5, FLJ21198, rapa-1, rapa-2, RP1-139D8.5, transcriptional regulating factor 1, transcriptional regulating protein 132, transcriptional-regulating factor 1, Transcriptional-regulating protein 132, TReP-132, TREP132, Zinc finger protein rapa, Zinc finger transcription factor TReP-132
Gene Symbols TRERF1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1150 and 1200 of human transcriptional regulating protein 132 (transcriptional regulating factor 1) using the numbering given in entry NP_277037.1 (GeneID 55809).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55809
Test Specificity Human. Based on 100% sequence identity, this antibody is predicted to react with Dog, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Small-eared galago, Thirteen-lined ground squirrel and Northern white-cheeked gibbon.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.