Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TREX1 Antibody (41M5F2) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB014065 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB014065 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB014065 Supplier Novus Biologicals Supplier No. NBP280992
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TREX1 Monoclonal specifically detects TREX1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TREX1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 41M5F2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 2-5 μg/mL, Flow Cytometry 2-5 ug/million cells/ml, Immunohistochemistry : 1:10-1:500, Immunohistochemistry-Paraffin : 5 μg/mL
Formulation PBS
Gene Alias 3'-5' exonuclease TREX1, AGS1, Aicardi-Goutieres syndrome 1, CRV, DKFZp434J0310, DNase III, DRN3, EC 3.1.11.2, HERNS, three prime repair exonuclease 1,3' repair exonuclease 1
Gene Symbols TREX1
Host Species Mouse
Immunogen Within the range of amino acids 211-350 of human TREX1 were used as immunogen for the antibody.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline DNA Repair, Editing and Processing Endonucleases
Primary or Secondary Primary
Gene ID (Entrez) 11277
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.