Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRF-1 Antibody (3H11), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB310282 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310282 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310282 Supplier Novus Biologicals Supplier No. NB12014397MFV610

Mouse Monoclonal Antibody

TRF-1 Monoclonal antibody specifically detects TRF-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen TRF-1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 3H11
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias NIMA-interacting protein 2, PIN2hTRF1-AS, Telomeric protein Pin2/TRF1, telomeric repeat binding factor (NIMA-interacting) 1, telomeric repeat binding factor 1, telomeric repeat binding protein 1, telomeric repeat-binding factor 1, TERF1, TRBF1, TRF1FLJ41416, TRFt-TRF1, TTAGGG repeat-binding factor 1
Host Species Mouse
Immunogen Human TRF-1
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7013
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.