Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRF-1 Antibody (57-6), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N1068281A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1068281A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1068281A70 Supplier Novus Biologicals Supplier No. NB11068281AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRF-1 Monoclonal specifically detects TRF-1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen TRF-1
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 57-6
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias NIMA-interacting protein 2, PIN2hTRF1-AS, Telomeric protein Pin2/TRF1, telomeric repeat binding factor (NIMA-interacting) 1, telomeric repeat binding factor 1, telomeric repeat binding protein 1, telomeric repeat-binding factor 1, TERF1, TRBF1, TRF1FLJ41416, TRFt-TRF1, TTAGGG repeat-binding factor 1
Gene Symbols TERF1
Host Species Mouse
Immunogen Recombinant human TRF1 [UniProtNo. P54274]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7013
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.