Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRF-1 Antibody (TRF-78) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB1001701 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001701 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001701 Supplier Novus Biologicals Supplier No. NB1001701
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

TRF-1 Monoclonal specifically detects TRF-1 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Microarray, Western Blot.

Specifications

Antigen TRF-1
Applications ELISA, Immunocytochemistry, Immunofluorescence, Microarray, Western Blot
Classification Monoclonal
Clone TRF-78
Concentration 2.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4 ug/ml, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Microarray
Gene Accession No. P54274
Gene Alias NIMA-interacting protein 2, PIN2hTRF1-AS, Telomeric protein Pin2/TRF1, telomeric repeat binding factor (NIMA-interacting) 1, telomeric repeat binding factor 1, telomeric repeat binding protein 1, telomeric repeat-binding factor 1, TERF1, TRBF1, TRF1FLJ41416, TRFt-TRF1, TTAGGG repeat-binding factor 1
Gene Symbols TERF1
Host Species Mouse
Immunogen Human TRF1 protein produced in baculovirus.
Molecular Weight of Antigen 70 kDa
Purification Method Protein G purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7013
Test Specificity TRF1 (TRF-78)
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.