Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRF-2 Antibody (4A794.15) - Azide Free, Novus Biologicals™
SDP

Catalog No. NBP233212 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP233212 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP233212 Supplier Novus Biologicals Supplier No. NBP233212
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRF-2 Monoclonal specifically detects TRF-2 in Human, Mouse, Rat, Deer, Mammal samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen TRF-2
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone 4A794.15
Concentration 0.5mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4 ug/ml, Simple Western, Flow Cytometry 1ul/1 million cells, ELISA 2 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 2 ug/10^6 cells, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 5 ug/ml, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Gene Accession No. Q15554
Gene Alias Telomeric DNA-binding protein, telomeric repeat binding factor 2, Telomeric repeat binding protein 2, telomeric repeat-binding factor 2, TERF 2, TERF2, TRBF 2, TRBF2, TRF 2, TRF2, TTAGGG repeat binding factor 2, TTAGGG repeat-binding factor 2
Gene Symbols TERF2
Host Species Mouse
Immunogen This TRF-2 Antibody (4A794.15) - Azide Free was developed against Baculovirus expressed whole length TRF2 protein, used for immunizing mice and splenocytes used to generate the hybridoma clone (NP_005643).
Molecular Weight of Antigen 59.6 kDa
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 7014
Test Specificity The recognizes distinct doublet bands of approximate size 66 kDa corresponding to the molecular size of huTRF2 by western blot analysis against lysates from most human cell lines.
Target Species Human, Mouse, Rat, Deer, Mammalia
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.