Learn More
Description
Specifications
Specifications
| Antigen | TRF-2 |
| Applications | Chemotaxis, ChIP Assay, CyTOF, ELISA, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunoblot, Immunocytochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Proximity Ligation Assay, Simple Western, Western Blot |
| Classification | Monoclonal |
| Clone | 4A794.15 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2-4 ug/ml, Simple Western 1:50, Flow Cytometry 0.1 ug/10^6 cells, ELISA 2 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 2 ug/10^6 cells, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 5 ug/ml, Immunoblotting reported in scientific literature (PMID 23708666), Proximity Ligation Assay reported in scientific literature (PMID 27366950), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, CyTOF-ready |
| Gene Accession No. | Q15554 |
| Gene Alias | 4A794, 4A794 trf2, 4A794.15, 4A794.15 trf2, anti-trf2 4A794, anti-trf2 4A794.15, clone 4A794, clone 4A794.15, Telomeric DNA-binding protein, telomeric repeat binding factor 2, Telomeric repeat binding protein 2, telomeric repeat-binding factor 2, TERF 2, TERF2, TRBF 2, TRBF2, TRF 2, TRF2, TTAGGG repeat binding factor 2, TTAGGG repeat-binding factor 2 |
| Gene Symbols | TERF2 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
