Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRF-2 Antibody (4A794.15) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $562.50
Specifications
Antigen | TRF-2 |
---|---|
Clone | 4A794.15 |
Concentration | 1 mg/mL |
Dilution | Western Blot 2-4 ug/ml, Simple Western 1:50, Flow Cytometry 0.1 ug/10^6 cells, ELISA 2 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 2 ug/10^6 cells, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 5 ug/ml, Immunoblotting reported in scientific literature (PMID 23708666), Proximity Ligation Assay reported in scientific literature (PMID 27366950), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056506T
![]() |
Novus Biologicals
NB10056506SS |
0.025 mg |
Each for $243.50
|
|
|||||
NB10056506
![]() |
Novus Biologicals
NB10056506 |
0.1 mg |
Each for $562.50
|
|
|||||
Description
TRF-2 Monoclonal specifically detects TRF-2 in Human, Mouse, Rat, Deer, Marsupial samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Proximity Ligation Assay, CyTOF-ready.Specifications
TRF-2 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Transcription Factors and Regulators | |
Q15554 | |
7014 | |
This TRF-2 Antibody (4A794.15) was developed against Baculovirus expressed whole length TRF2 protein, used for immunizing mice and splenocytes used to generate the hybridoma clone (NP_005643). | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
59.6 kDa |
4A794.15 | |
Western Blot 2-4 ug/ml, Simple Western 1:50, Flow Cytometry 0.1 ug/10^6 cells, ELISA 2 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 2 ug/10^6 cells, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 5 ug/ml, Immunoblotting reported in scientific literature (PMID 23708666), Proximity Ligation Assay reported in scientific literature (PMID 27366950), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Deer, Marsupial | |
4A794, 4A794 trf2, 4A794.15, 4A794.15 trf2, anti-trf2 4A794, anti-trf2 4A794.15, clone 4A794, clone 4A794.15, Telomeric DNA-binding protein, telomeric repeat binding factor 2, Telomeric repeat binding protein 2, telomeric repeat-binding factor 2, TERF 2, TERF2, TRBF 2, TRBF2, TRF 2, TRF2, TTAGGG repeat binding factor 2, TTAGGG repeat-binding factor 2 | |
TERF2 | |
IgG1 κ | |
Protein G purified | |
The TRF2 antibody recognizes full-length TRF2 as well as TRF2 forms lacking both the N-terminal basic domain (B) and the telobox Myb-like C-terminal DNA-binding domain (M). TRF2 forms missing the B and M domains are often referred to as mutant TRF2. Although the exact epitope recognized by the TRF2 antibody has not been mapped, the scientific literature indicates it is in the D or L domain, but not in the B or M domain. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title