Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRF-2 Antibody (4A794.15), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB10056506V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056506V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056506V Supplier Novus Biologicals Supplier No. NB10056506V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRF-2 Monoclonal specifically detects TRF-2 in Deer, Human, Marsupial, Mouse, Rat samples. It is validated for ChIP Assay, CyTOF, ELISA, Flow Cytometry, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Proximity Ligation Assay, Western Blot.

Specifications

Antigen TRF-2
Applications ChIP Assay, CyTOF, ELISA, Flow Cytometry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Proximity Ligation Assay, Western Blot
Classification Monoclonal
Clone 4A794.15
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Accession No. Q15554
Gene Alias Telomeric DNA-binding protein, telomeric repeat binding factor 2, Telomeric repeat binding protein 2, telomeric repeat-binding factor 2, TERF 2, TERF2, TRBF 2, TRBF2, TRF 2, TRF2, TTAGGG repeat binding factor 2, TTAGGG repeat-binding factor 2
Gene Symbols TERF2
Host Species Mouse
Immunogen This TRF-2 Antibody (4A794.15) [DyLight 405] was developed against Baculovirus expressed whole length TRF2 protein, used for immunizing mice and splenocytes used to generate the hybridoma clone (NP_005643).
Molecular Weight of Antigen 59.6 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 7014
Test Specificity The TRF2 antibody recognizes full-length TRF2 as well as TRF2 forms lacking both the N-terminal basic domain (B) and the telobox Myb-like C-terminal DNA-binding domain (M). TRF2 forms missing the B and M domains are often referred to as mutant TRF2. Although the exact epitope recognized by the TRF2 antibody has not been mapped, the scientific literature indicates it is in the D or L domain, but not in the B or M domain.
Target Species Deer, Human, Marsupial, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.