Learn More
Description
Specifications
Specifications
| Antigen | TRF-2 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Dot Blot, KnockDown |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000 - 1:5000, Simple Western 1:25, Flow Cytometry 1-5 ug/ml, ELISA reported in scientific literature (PMID 31575660), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature, Immunohistochemistry-Paraffin 1:200, Dot Blot reported in scientific literature (PMID 31026066), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, KnockDown Validated reported in scientific literature (PMID 31026066) |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | Q15554 |
| Gene Alias | Telomeric DNA-binding protein, telomeric repeat binding factor 2, Telomeric repeat binding protein 2, telomeric repeat-binding factor 2, TERF 2, TERF2, TRBF 2, TRBF2, TRF 2, TRF2, TTAGGG repeat binding factor 2, TTAGGG repeat-binding factor 2 |
| Gene Symbols | TERF2 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
