Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRF-2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11057130T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11057130T 0.025 mL
NB11057130 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11057130T Supplier Novus Biologicals Supplier No. NB11057130SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 86 publications

TRF-2 Polyclonal specifically detects TRF-2 in Human, Mouse, Rat, Chinese Hamster, Primate samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Dot Blot, Knockdown Validated.

Specifications

Antigen TRF-2
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Dot Blot, KnockDown
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:5000, Simple Western 1:25, Flow Cytometry 1-5 ug/ml, ELISA reported in scientific literature (PMID 31575660), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature, Immunohistochemistry-Paraffin 1:200, Dot Blot reported in scientific literature (PMID 31026066), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, KnockDown Validated reported in scientific literature (PMID 31026066)
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q15554
Gene Alias Telomeric DNA-binding protein, telomeric repeat binding factor 2, Telomeric repeat binding protein 2, telomeric repeat-binding factor 2, TERF 2, TERF2, TRBF 2, TRBF2, TRF 2, TRF2, TTAGGG repeat binding factor 2, TTAGGG repeat-binding factor 2
Gene Symbols TERF2
Host Species Rabbit
Immunogen This TRF-2 Antibody was developed against Baculovirus purified TRF2 protein.
Molecular Weight of Antigen 59.6 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 7014
Target Species Human, Mouse, Rat, Chinese Hamster, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.