Learn More
Tri(ethylene glycol) di-p-toluenesulfonate, 98%
Description
Tri(ethylene glycol) di-p-toluenesulfonate is used in the synthesis of 1-[4-(1,4,7,10-tetraoxa-13-aza-cyclopentadec-13-yl)-phenyl] ethanone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tri(ethylene glycol) di-p-toluenesulfonate is used in the synthesis of 1-[4-(1,4,7,10-tetraoxa-13-aza-cyclopentadec-13-yl)-phenyl] ethanone.
Solubility
Soluble in ethanol.
Notes
Incompatible with strong bases.
Specifications
Specifications
| Melting Point | 80°C to 82°C |
| Quantity | 50 g |
| Solubility Information | Soluble in ethanol. |
| Formula Weight | 458.55 |
| Percent Purity | 98% |
| Chemical Name or Material | Tri(ethylene glycol) di-p-toluenesulfonate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.