Learn More
Tri-n-butyltin hydride, 97%
Description
Tri-n-butyltin hydride is used as a radical reagent in reductive cleavage, radical dehalogenation and intramolecular radical cyclization. It also promotes intramolecular cyclization leading to isoxazolo-benzaulene ring system. Further, it acts as a reducing agent for the conversion of alkyl halides to hydrocarbons. In addition to this, it is used in a veterinary anthelmintic for poultry and an intermediate to make other butyltin compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tri-n-butyltin hydride is used as a radical reagent in reductive cleavage, radical dehalogenation and intramolecular radical cyclization. It also promotes intramolecular cyclization leading to isoxazolo-benzaulene ring system. Further, it acts as a reducing agent for the conversion of alkyl halides to hydrocarbons. In addition to this, it is used in a veterinary anthelmintic for poultry and an intermediate to make other butyltin compounds.
Solubility
Immiscible with water.
Notes
Air and moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and acids.
Specifications
Specifications
| Melting Point | 0°C to 1°C |
| Density | 1.098 |
| Boiling Point | 68°C to 74°C (0.3 mmHg) |
| Flash Point | 40°C (104°F) |
| Odor | Odorless |
| Refractive Index | 1.473 |
| Quantity | 10 g |
| UN Number | UN2929 |
| Beilstein | 3587329 |
| Sensitivity | Air sensitive; May form a precipitate |
| Show More |
Safety and Handling
| Hazard Category | H301-H312-H315-H317-H319-H360FD-H372 |
| Hazard Statement | GHS H Statement H301-H372-H226-H312-H315-H319 Toxic if swallowed. Causes damage to organs through prolonged or repeated exposure. Flammable liquid and vapour. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S. |
| EINECSNumber | 211-704-4 |
| RTECSNumber | WH8675000 |
| TSCA | Yes |
| Recommended Storage | Keep cold; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.