Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-n-butyltin hydride, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
688-73-3
C12H28Sn
291.04
MFCD00009416
PIILXFBHQILWPS-UHFFFAOYSA-N
tributylstannanyl,tributlytin,tributylstannic hydride,tributyl tin,tri-n-butylstannane hydride,unii-4xdx163p3d,tin, tri-n-butyl-, hydride,tri-n-butylstannyl radical,stannane, tri-n-butyl-, hydride,tributyl-tin
3032732
tributyltin
CCCC[Sn](CCCC)CCCC
688-73-3
C12H28Sn
291.04
MFCD00009416
PIILXFBHQILWPS-UHFFFAOYSA-N
tributylstannanyl,tributlytin,tributylstannic hydride,tributyl tin,tri-n-butylstannane hydride,unii-4xdx163p3d,tin, tri-n-butyl-, hydride,tri-n-butylstannyl radical,stannane, tri-n-butyl-, hydride,tributyl-tin
3032732
tributyltin
CCCC[Sn](CCCC)CCCC
Reduction reagent
Quantity 10g
Packaging Glass bottle
Density 1.0800g/mL
Specific Gravity 1.08
Infrared Spectrum Authentic
Boiling Point 80.0°C (0.4 mmHg)
Refractive Index 1.4720 to 1.4740
Color Colorless
Viscosity 1.5 mPa.s (20°C)
Flash Point 40°C
Assay Percent Range 96% min. (GC)
Linear Formula [CH3(CH2)3]3SnH
Fieser 01,1192; 02,424; 03,294; 04,518; 05,685; 06,604; 07,379; 08,497; 09,476; 10,411; 11,545; 12,516; 14,312; 15,325; 16,343; 17,351
Merck Index 15, 9783
Solubility Information Solubility in water: reacts. Other solubilities: soluble in most organic solvents
Formula Weight 291.04
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Tri-n-butyltin hydride
Show More Show Less

GHS H Statement
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Causes damage to organs through prolonged

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiou

GHS Signal Word: Danger

EINECSNumber : 211-704-4