Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-o-cresyl Phosphate 97.0+%, TCI America™
SDP

Catalog No. P133125G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Catalog No. P133125G

Supplier: TCI America P133125G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 78-30-8
Molecular Formula C21H21O4P
Molecular Weight (g/mol) 368.37
MDL Number MFCD00041906
InChI Key YSMRWXYRXBRSND-UHFFFAOYSA-N
Synonym tri-o-cresyl phosphate, tri-o-tolyl phosphate, o-tolyl phosphate, tocp, totp, phosphoric acid, tris 2-methylphenyl ester, o-cresyl phosphate, triorthocresyl phosphate, phosflex 179c, tri-2-tolyl phosphate
PubChem CID 6527
IUPAC Name tris(2-methylphenyl) phosphate
SMILES CC1=CC=CC=C1OP(=O)(OC1=CC=CC=C1C)OC1=CC=CC=C1C

Specifications

Color Yellow
Boiling Point 410°C
Quantity 25 g
UN Number 2574
Formula Weight 368.37
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tri-o-cresyl Phosphate
EINECSNumber (3)-2522&(3)-3363&(3)-2613
RTECSNumber TD0350000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.