Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triacontane, 98%

Catalog No. p-3758846
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 638-68-6 | C30H62 | 422.826 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-68-6
MDL Number MFCD00009410
InChI Key JXTPJDDICSTXJX-UHFFFAOYSA-N
Synonym n-triacontane, triacontane, analytical standard, triacontane, acmc-1b4uf, n-triacontane 250mg, ch3-ch2 28-ch3
PubChem CID 12535
ChEBI CHEBI:31006
IUPAC Name triacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 65°C to 67°C
Color White
Boiling Point 455°C
Flash Point 238°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C30H62
Linear Formula CH3(CH2)28CH3
Quantity 1 g
Beilstein 01, 176
Solubility Information Solubility in water: insoluble
Molecular Weight (g/mol) 422.82
Formula Weight 422.82
Percent Purity 98%
Physical Form Shiny Flakes
Chemical Name or Material Triacontane
Show More Show Less
EINECSNumber 211-349-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.