Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIAD3 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NB10056891

 View more versions of this product

Catalog No. NB10056891


Only null left
Add to Cart

Description

Description

TRIAD3 Polyclonal specifically detects TRIAD3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

TRIAD3
Polyclonal
Unconjugated
PBS, 0.2% gelatin with 0.05% Sodium Azide
EC 6.3.2.-, ring finger protein 216Ubiquitin-conjugating enzyme 7-interacting protein 1, Triad domain-containing protein 3, TRIAD3Zinc finger protein inhibiting NF-kappa-B, U7I1, UBCE7IP1ubiquitin conjugating enzyme 7 interacting protein 1, ZINE3 ubiquitin-protein ligase RNF216
Rabbit
Protein G purified
RUO
54476
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot
0.5 mg/mL
Western Blot 1 - 3 ug/mL
NP_996999
RNF216
This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human TRIAD3 between amino acids 350-400.
0.1 mg
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only