Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIAD3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | TRIAD3 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1 - 3 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056891T
![]() |
Novus Biologicals
NB10056891SS |
0.025 mg |
Each for $181.90
|
|
|||||
NB10056891
![]() |
Novus Biologicals
NB10056891 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
TRIAD3 Polyclonal specifically detects TRIAD3 in Human samples. It is validated for Western Blot.Specifications
TRIAD3 | |
Western Blot 1 - 3 ug/mL | |
Polyclonal | |
Purified | |
RUO | |
PBS, 0.2% gelatin with 0.05% Sodium Azide | |
EC 6.3.2.-, ring finger protein 216Ubiquitin-conjugating enzyme 7-interacting protein 1, Triad domain-containing protein 3, TRIAD3Zinc finger protein inhibiting NF-kappa-B, U7I1, UBCE7IP1ubiquitin conjugating enzyme 7 interacting protein 1, ZINE3 ubiquitin-protein ligase RNF216 | |
RNF216 | |
IgG | |
Protein G purified |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
NP_996999 | |
54476 | |
This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human TRIAD3 between amino acids 350-400. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title