Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIAD3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10056891SS
Description
TRIAD3 Polyclonal specifically detects TRIAD3 in Human samples. It is validated for Western Blot.Specifications
TRIAD3 | |
Polyclonal | |
Unconjugated | |
PBS, 0.2% gelatin with 0.05% Sodium Azide | |
EC 6.3.2.-, ring finger protein 216Ubiquitin-conjugating enzyme 7-interacting protein 1, Triad domain-containing protein 3, TRIAD3Zinc finger protein inhibiting NF-kappa-B, U7I1, UBCE7IP1ubiquitin conjugating enzyme 7 interacting protein 1, ZINE3 ubiquitin-protein ligase RNF216 | |
Rabbit | |
Protein G purified | |
RUO | |
54476 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
0.5 mg/mL | |
Western Blot 1 - 3 ug/mL | |
NP_996999 | |
RNF216 | |
This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of human TRIAD3 between amino acids 350-400. | |
0.025 mg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction