Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIB1 Antibody (OTI8C8), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP27496A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27496A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27496A32 Supplier Novus Biologicals Supplier No. NBP274596AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRIB1 Monoclonal specifically detects TRIB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen TRIB1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI8C8
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias C8FWG-protein-coupled receptor induced protein, GIG-2, GIG2G-protein-coupled receptor-induced protein 2, G-protein-coupled receptor-induced gene 2 protein, SKIP1, TRB-1, TRB1phosphoprotein regulated by mitogenic pathways, tribbles homolog 1, tribbles homolog 1 (Drosophila), tribbles-like protein 1
Gene Symbols TRIB1
Host Species Mouse
Immunogen Full length recombinant protein of human TRIB1 (NP_079471) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 10221.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.