Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIB3 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB184541 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB184541 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB184541 Supplier Novus Biologicals Supplier No. NBP282067AF350
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRIB3 Polyclonal antibody specifically detects TRIB3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen TRIB3
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias C20orf97, dJ1103G7.3, neuronal cell death inducible putative kinase, Neuronal cell death-inducible putative kinase, NIPK, p65-interacting inhibitor of NF-kappaB, p65-interacting inhibitor of NF-kappa-B, SINK, SKIP3, TRB-3, TRB3chromosome 20 open reading frame 97, tribbles homolog 3, tribbles homolog 3 (Drosophila)
Host Species Rabbit
Immunogen TRIB3 antibody was raised against a 17 amino acid peptide near the center of human TRIB3. The immunogen is located within amino acids 100 - 150 of TRIB3. Amino Acid Squence: EPYARLPPHKHVARPTE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 57761
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.