Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIB3 Mouse anti-Human, Mouse, Rat, Clone: 2C10C7, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | TRIB3 |
---|---|
Clone | 2C10C7 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7118
![]() ![]() |
Proteintech Group Inc
667021IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
A novel inhibitor of cell division, Tribbles, blocks mitosis at a critical point in Drosophila morphogenesis. Recently, mammalian homologues of Tribbles (Trbs) have been identified that control mitogen-activated protein kinase (MAPK) activity. TRIB3 also known as TRB-3, SINK, and SKIP3 is a member of Trbs family. Overexpression of this protein inhibits NF-kB-dependent transcription induced by tumor necrosis factor (TNF) stimulation or its downstream signaling proteins but does not inhibit NF-kB translocation to the nucleus and binding to DNA. TRIB3/SKIP3 is also overexpressed in multiple human tumors and is regulated by hypoxia.Specifications
TRIB3 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C20orf97, NIPK, SINK, SKIP3, TRB 3, TRB3, TRIB3, Tribbles homolog 3 | |
TRIB3 | |
IgG1 | |
Protein G | |
Antibody |
2C10C7 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
Q8K4K2, Q96RU7, Q9WTQ6 | |
228775, 246273, 57761 | |
TRIB3 Fusion Protein Ag25776 | |
Primary | |
-20°C | |
TRIB3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title