Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIB3 Rabbit anti-Human, Clone: S01-9D1, Novus Biologicals™
SDP

Catalog No. NB165413 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165413 100 μg
NB165414 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB165413 Supplier Novus Biologicals Supplier No. NBP315103100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TRIB3 Monoclonal antibody specifically detects TRIB3 in Human samples. It is validated for Western Blot

Specifications

Antigen TRIB3
Applications Western Blot
Classification Monoclonal
Clone S01-9D1
Conjugate Unconjugated
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias C20orf97, dJ1103G7.3, neuronal cell death inducible putative kinase, Neuronal cell death-inducible putative kinase, NIPK, p65-interacting inhibitor of NF-kappaB, p65-interacting inhibitor of NF-kappa-B, SINK, SKIP3, TRB-3, TRB3chromosome 20 open reading frame 97, tribbles homolog 3, tribbles homolog 3 (Drosophila)
Host Species Rabbit
Immunogen A synthetic peptide of human TRIB3 (Uniprot#: Q96RU7)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 57761
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.