Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tricarballylic acid, 98%

Catalog No. AAB2210314
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAB2210314

Supplier: Thermo Scientific Chemicals B2210314

Only null left
Add to Cart

CAS: 99-14-9 | C6H8O6 | 176.12 g/mol

It is used as an alternative to DMDHEU (formaldehyde releasing cross- linking agent).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an alternative to DMDHEU (formaldehyde releasing cross- linking agent).

Solubility
Soluble in water (500 mg/ml at 18°C), methanol, and ether (slightly).

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-14-9
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00002723
InChI Key KQTIIICEAUMSDG-UHFFFAOYSA-N
Synonym tricarballylic acid, 1,2,3-propanetricarboxylic acid, carballylic acid, tricarballylate, beta-carboxyglutaric acid, propane 1,2,3-tricarboxylic acid, unii-ra5qh2j020, 3-carboxyglutaric acid, .beta.-carboxyglutaric acid, 1,2,3-propanetricarboxylicacid
PubChem CID 14925
ChEBI CHEBI:45969
IUPAC Name propane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(CC(O)=O)C(O)=O

Specifications

Melting Point 157°C to 162°C
Quantity 25 g
Beilstein 1783567
Merck Index 14,9622
Solubility Information Soluble in water (500mg/ml at 18°C),methanol,and ether (slightly).
Formula Weight 176.12
Percent Purity 98%
Chemical Name or Material Tricarballylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-733-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.