Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Manganese(II) acetate tetrahydrate, 99+%, for analysis
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
| Melting Point | >300.0°C |
| Color | Pink |
| pH | 6 to 7 (5% soln.) |
| Physical Form | Crystals |
| Quantity | 2.5 kg |
| Assay Percent Range | 99% min. (Complexometry) |
| Linear Formula | (CH3CO2)2Mn·4H2O |
| Flash Point | >130°C |
| Merck Index | 14, 5724 |
| Solubility Information | Solubility in water: very soluble. Other solubilities: soluble in alcohol |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Con |
| RTECSNumber | AI5775000 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction