Learn More
Trichloroethylene, ACS, 99.5% min
Description
Trichloroethylene is used as solvent for paints, dyes, waxes, oils, fats and resins, as an additive in polymerization reactions, as a reactant to prepare various halohydrocarbons (1,1,1,2-tetrafluoroethane) and other chemicals, as a coolant for cutting tantalum and acts as good heat transfer medium. It is used in fire extinguisher fluids and in the recovery of oils and greases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 85°C |
| Density | 1.46 |
| Boiling Point | 87°C |
| Flash Point | None |
| Assay Percent Range | 9.5% min. |
| Refractive Index | 1.476 |
| Quantity | 500 mL |
| UN Number | UN1710 |
| Beilstein | 1736782 |
| Merck Index | 14,9639 |
| Show More |
Safety and Handling
| Hazard Category | H315-H317-H319-H336-H341-H350-H373 |
| Hazard Statement | GHS H Statement H350-H341-H315-H319-H336 |
| Precautionary Statement | P201-P202-P260-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TRICHLOROETHYLENE |
| EINECSNumber | 201-167-4 |
| RTECSNumber | KX4550000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.