Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroethylene, ACS, 99.5% min

Catalog No. AA19401K2
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
This item is not returnable. View return policy

Catalog No. AA19401K2

Supplier: Thermo Scientific Chemicals 019401K2

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 79-01-6 | C2HCl3 | 131.38 g/mol

Trichloroethylene is used as solvent for paints, dyes, waxes, oils, fats and resins, as an additive in polymerization reactions, as a reactant to prepare various halohydrocarbons (1,1,1,2-tetrafluoroethane) and other chemicals, as a coolant for cutting tantalum and acts as good heat transfer medium. It is used in fire extinguisher fluids and in the recovery of oils and greases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-01-6
Molecular Formula C2HCl3
Molecular Weight (g/mol) 131.38
MDL Number MFCD00000838
InChI Key XSTXAVWGXDQKEL-UHFFFAOYSA-N
Synonym trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
PubChem CID 6575
ChEBI CHEBI:16602
IUPAC Name 1,1,2-trichloroethene
SMILES C(=C(Cl)Cl)Cl

Specifications

Melting Point -85°C
Density 1.46 g/mL
Boiling Point 87°C
Flash Point None
Odor Chloroform-like
Assay Percent Range 9.5% min.
Refractive Index 1.476
Quantity 1 L
UN Number UN1710
Beilstein 1736782
Merck Index 14,9639
Solubility Information Miscible with ether,alcohol,chloroformMiscible with ether,ethanol and chloroform; Very slightly miscible with water.
Formula Weight 131.39
Percent Purity ≤99.5%
Grade ACS
Physical Form Liquid
Chemical Name or Material Trichloroethylene, For ACS analysis
Show More Show Less
Hazard Category H315-H317-H319-H336-H341-H350-H373
Hazard Statement GHS H Statement
H350-H341-H315-H319-H336
May cause cancer.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P260-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TRICHLOROETHYLENE
EINECSNumber 201-167-4
RTECSNumber KX4550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.