Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trichloroisocyanuric acid, 90+%
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
Trichloroisocyanuric acid was used as a disinfection reagent in the transformation of benzophenone-3 in chlorinated water. It was also used in the synthesis of 2-halomethylene-3-oxoketoximes. It is used as a bleaching agent in the textile industry, a reagent in organic synthesis.
Solubility
Soluble in chlorocarbon, acetonitrile, acetone, water (12 g/L).
Notes
Moisture sensitive. Store away from reducing agents, acids, bases, flammable substances, organic materials, light, water/ moisture.
Trichloroisocyanuric acid was used as a disinfection reagent in the transformation of benzophenone-3 in chlorinated water. It was also used in the synthesis of 2-halomethylene-3-oxoketoximes. It is used as a bleaching agent in the textile industry, a reagent in organic synthesis.
Solubility
Soluble in chlorocarbon, acetonitrile, acetone, water (12 g/L).
Notes
Moisture sensitive. Store away from reducing agents, acids, bases, flammable substances, organic materials, light, water/ moisture.
Specifications
Specifications
| Melting Point | ∼245°C (decomposition) |
| Density | 2.07 |
| pH | 3 |
| Flash Point | >250°C (482°F) |
| Odor | Pungent |
| Quantity | 2500 g |
| UN Number | UN2468 |
| Beilstein | 202022 |
| Sensitivity | Moisture sensitive |
| Merck Index | 14,9641 |
| Show More |
Safety and Handling
| Hazard Category | H272-H302-H319-H335 |
| Hazard Statement | GHS H Statement H272-H302-H319-H335 May intensify fire; oxidizer. Harmful if swallowed. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P312-P330-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: TRICHLOROISOCYANURIC ACID, DRY |
| EINECSNumber | 201-782-8 |
| RTECSNumber | XZ1925000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction