Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloromethanesulfonyl Chloride 95.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
2547-61-7
CCl4O2S
217.869
MFCD00007452
ZCPSWAFANXCCOT-UHFFFAOYSA-N
75684
trichloromethanesulfonyl chloride
C(S(=O)(=O)Cl)(Cl)(Cl)Cl
2547-61-7
CCl4O2S
217.869
MFCD00007452
ZCPSWAFANXCCOT-UHFFFAOYSA-N
75684
trichloromethanesulfonyl chloride
C(S(=O)(=O)Cl)(Cl)(Cl)Cl
Color White-Yellow
Melting Point 141°C
Quantity 25g
UN Number 3261
Formula Weight 217.87
Physical Form Crystal-Powder at 20°C
Percent Purity ≥95.0% (T)
Chemical Name or Material Trichloromethanesulfonyl Chloride