Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichomonas Vaginalis 1 Antibody (1306/156), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N1062964A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1062964A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1062964A70 Supplier Novus Biologicals Supplier No. NB10062964AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Trichomonas Vaginalis 1 Monoclonal specifically detects Trichomonas Vaginalis 1 in Protozoa samples. It is validated for ELISA.

Specifications

Antigen Trichomonas Vaginalis 1
Applications ELISA
Classification Monoclonal
Clone 1306/156
Conjugate Alexa Fluor 750
Dilution ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias T vaginalis, TV
Host Species Mouse
Immunogen Native
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Recognises T. vaginalis strain 1, an anaerobic parasitic protozoan and causative agent of the sexually transmitted disease Trichomoniasis.
Target Species Protozoa
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.