Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichomonas Vaginalis 1 Antibody (1306/156), HRP, Novus Biologicals™
SDP

Catalog No. NB10062964H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062964H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062964H Supplier Novus Biologicals Supplier No. NB10062964H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Trichomonas Vaginalis 1 Monoclonal specifically detects Trichomonas Vaginalis 1 in Protozoa samples. It is validated for ELISA.

Specifications

Antigen Trichomonas Vaginalis 1
Applications ELISA
Classification Monoclonal
Clone 1306/156
Conjugate HRP
Dilution ELISA
Formulation PBS with No Preservative
Gene Alias T vaginalis, TV
Host Species Mouse
Immunogen Native
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognises T. vaginalis strain 1, an anaerobic parasitic protozoan and causative agent of the sexually transmitted disease Trichomoniasis.
Target Species Protozoa
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.