Learn More
Tricyclohexyltin chloride, 97%
Description
Tricyclohexyltin chloride is used in the preparation of dicyclohexyl disulfide. It is also used in the synthesis of organotin derivatives of 2-thiophene carboxylic acid as enzyme inhibitors and antibacterial agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tricyclohexyltin chloride is used in the preparation of dicyclohexyl disulfide. It is also used in the synthesis of organotin derivatives of 2-thiophene carboxylic acid as enzyme inhibitors and antibacterial agents.
Solubility
Soluble in ethanol, ether, toluene and chloroform. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 123°C to 125°C |
| Linear Formula | (C6H11)3SnCl |
| Quantity | 1 g |
| UN Number | UN3146 |
| Solubility Information | Soluble in ethanol,ether,toluene and chloroform. Insoluble in water. |
| Formula Weight | 403.6 |
| Percent Purity | 97% |
| Chemical Name or Material | Tricyclohexyltin chloride |
Safety and Handling
| Hazard Category | H302+H312+H332 |
| Hazard Statement | GHS H Statement H302-H312-H332 Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S. |
| EINECSNumber | 221-437-5 |
| RTECSNumber | WH6830000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.