Learn More
Triethyl 2-phosphonobutyrate, 97%
Description
Reactant used for preparation of furospinosulin-1 and analogs as hypoxia-selective antitumor agents, Aminoquinolines as beta-site amyloid precursor protein cleaving enzyme 1 (BACE1) inhibitors and potential anti-Alzheimer?s drugs, phenylpropanoic acid peroxisome proliferator-activated receptor (PPAR) α-selective agonists as nonalcoholic steatohepatitis (NASH)-preventive agents, PPAR agonists with phenethylphenylphthalimide skeleton derived from thalidomide-related LXR antagonists, notch-sparing γ-secretase inhibitors derived from PPAR agonist library and plakotenin via Diels-Alder reaction, as a potential anticancer agent, naturally found in Okinawan sponge of the genus Plakortis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reactant used for preparation of furospinosulin-1 and analogs as hypoxia-selective antitumor agents, Aminoquinolines as beta-site amyloid precursor protein cleaving enzyme 1 (BACE1) inhibitors and potential anti-Alzheimer′s drugs, phenylpropanoic acid peroxisome proliferator-activated receptor (PPAR) α-selective agonists as nonalcoholic steatohepatitis (NASH)-preventive agents, PPAR agonists with phenethylphenylphthalimide skeleton derived from thalidomide-related LXR antagonists, notch-sparing γ-secretase inhibitors derived from PPAR agonist library and plakotenin via Diels-Alder reaction, as a potential anticancer agent, naturally found in Okinawan sponge of the genus Plakortis.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
| Density | 1.064 |
| Boiling Point | 105°C to 106°C (2 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.433 |
| Quantity | 5 g |
| Beilstein | 1789280 |
| Solubility Information | Not miscible or difficult to mix with water. |
| Formula Weight | 252.25 |
| Percent Purity | 97% |
| Chemical Name or Material | Triethyl 2-phosphonobutyrate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.