Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl orthoacetate 97%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
2.5L
78-39-7
C8H18O3
162.23
MFCD00009223
NDQXKKFRNOPRDW-UHFFFAOYSA-N
triethyl orthoacetate,ethyl orthoacetate,ethane, 1,1,1-triethoxy,orthoacetic acid, triethyl ester,orthoacetic acid triethyl ester,triethoxyethane,triethoxy ethane,triethy orthoacetate,triethyl ortho-aceate,1,1-triethoxyethane
66221
1,1,1-triethoxyethane
CCOC(C)(OCC)OCC
78-39-7
C8H18O3
162.23
MFCD00009223
NDQXKKFRNOPRDW-UHFFFAOYSA-N
triethyl orthoacetate,ethyl orthoacetate,ethane, 1,1,1-triethoxy,orthoacetic acid, triethyl ester,orthoacetic acid triethyl ester,triethoxyethane,triethoxy ethane,triethy orthoacetate,triethyl ortho-aceate,1,1-triethoxyethane
66221
1,1,1-triethoxyethane
CCOC(C)(OCC)OCC
Viscosity 1 mPa.s (20°C)
Quantity 100mL
Refractive Index 1.3950 to 1.3980
Specific Gravity 0.885
Density 0.8850g/mL
Infrared Spectrum Authentic
Color Colorless to Yellow
Flash Point 36°C
Boiling Point 142.0°C
Packaging Glass bottle
Assay Percent Range 97%
Linear Formula CH3C(OC2H5)3
Beilstein 02, 129
Fieser 03,300; 04,527; 06,607; 10,417
Solubility Information Solubility in water: decomposes
Formula Weight 162.23
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Triethyl orthoacetate, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 201-112-4

TSCA : TSCA